Learn More
Cytiva (Formerly Pall Lab) AcroPrep™ 96 Filter Plates, 1 mL - 1.0 m, glass fiber media (5/pkg)
Supplier:  Cytiva 5051N
Encompass_Preferred
Optimized for a Variety of Applications Requiring Larger Per Well Volume ProcessingPlates are constructed of chemically resistant, biologically-inert polypropyleneRobotics-friendly design has single piece construction, designed in accordance with the ANSI/SBS X-2004 specificationsA serialized barcode label allows use in automated tracking systemsProprietary sealing technology individually seals each well, eliminating crosstalkAvailable in a variety of membrane configurationsExtended flow directors eliminate splattering and crosstalk of filtrate This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-206-3127
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.