Learn More
Cytiva AcroPrep™ 384-well Filter Plates, 100 µL - 30K Omega membrane, short tips, natural (10/pkg)
Supplier:  Cytiva 5078
Encompass_Preferred
Ideal for Use With Low Sample Volume, High Throughput Sample Prep Applications-Plates are constructed from chemically-resistant, biologically-inert polypropylene with a clear polystyrene lid.-Robotic-friendly design has single-piece construction, designed in accordance with the ANSI/SBS X-2004 standards. Plates are stackable with and without lids.-Eliminates crosstalk between wells. Membranes are individually sealed in each well with a proprietary sealing process and each well has a fluid director.-A serialized barcode label allows the use of automated tracking systems and identifies the membrane type.-Available in a variety of membrane configurations to meet the requirements of numerous applications. This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-206-3132
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.