Learn More
Thermo Scientific™ AcroMetrix™ Controls (BB NAT)
Assess the analytical performance of NAT test procedures for the qualitative determination of viral nucleic acids with these controls, which are for In Vitro Diagnostic Use.
Supplier: Thermo Scientific™ 963602
Description
Assess the analytical performance of molecular diagnostic test procedures for the qualitative determination of viral nucleic acids with the Thermo Scientific™ AcroMetrix™ Controls. These controls are intended for use with the detection of viral nucleic acids from donations of whole blood and blood components for transfusion. The controls are for In Vitro Diagnostic Use.
Specifications
AcroMetrix BB NAT HIV-1 Positive Control | |
Plasma, Serum | |
10 x 1.4mL |
HIV-1 | |
Control | |
10 x 1.4 mL |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.