Learn More
Thermo Scientific™ AcroMetrix™ MultiMix FFPE Control (RUO)
Assess performance of somatic genetic variant detection in research oncology testing with these controls, which are for Research Use Only.
Supplier: Thermo Scientific™ 957185
Description
Assess the performance of methods for the isolation, purification, and detection of somatic genetic variants in research oncology testing with the Thermo Scientific™ AcroMetrix™ MultiMix FFPE Controls (RUO). These controls consist of five single-use samples of a synthetic tissue that has been formalin-fixed, paraffin-embedded and sectioned to 10 microns thick. The controls are for Research Use Only and are not for use in diagnostic procedures.
Specifications
MultiMix B | |
MultiMix FFPE Control | |
5 samples | |
FBXW7, FLT3, RET, KRAS, PIK3CA, APC, TP53, FGFR3, KDR, PDGFRA, EGFR |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.