Learn More
Thermo Scientific™ AcroMetrix™ MultiMix FFPE Control (RUO)
Assess performance of somatic genetic variant detection in research oncology testing with these controls, which are for Research Use Only.
Supplier:  Thermo Scientific™ 957184
 View more versions of this product
Catalog No. 23-279-196
Description
Description
Assess the performance of methods for the isolation, purification, and detection of somatic genetic variants in research oncology testing with the Thermo Scientific™ AcroMetrix™ MultiMix FFPE Controls (RUO). These controls consist of five single-use samples of a synthetic tissue that has been formalin-fixed, paraffin-embedded and sectioned to 10 microns thick. The controls are for Research Use Only and are not for use in diagnostic procedures.
Specifications
Specifications
MultiMix A
MultiMix FFPE Control
5 samples
RET, EGFR, CSF1R, PDGFRA, GNA11, HRAS, TP53, FLT3, FBXW7, HNF1A, VHL, FGFR3, PIK3CA, STK11
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.