Learn More
Thermo Scientific™ AcroMetrix™ HIV-1/HCV/HBV Panels (RUO)

Assess the performance of molecular test procedures for HIV-1, HCV and HBV nucleic acids with these panels, which are for Research Use Only.
$1348.00 - $2899.00
Catalog Number | Mfr. No. | Description | Level | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Level | Price | Quantity & Availability | ||||
23-279-113
|
Thermo Scientific™
942013 |
AcroMetrix HIV-1 Panel (IU/mL) | 0, 100, 1000, 10000, 100000, 1000000, 10000000 IU/mL |
Pack of 7 for $1,348.00
|
|
||||
23-279-114
![]() |
Thermo Scientific™
950470 |
AcroMetrix HIV-1 Panel (copies/mL) | 0, 100, 500, 5000, 50000, 500000, 5000000 copies/mL |
Pack of 7 for $1,446.00
|
|
||||
23-279-123
![]() |
Thermo Scientific™
950350 |
AcroMetrix HCV-s Panel | 100, 500, 5000, 50000, 500000, 5000000, 25000000 IU/mL |
Pack of 7 for $2,899.00
|
|
||||
23-279-119
![]() |
Thermo Scientific™
950150 |
AcroMetrix HBV Panel | 50, 500, 5000, 50000, 500000, 5000000, 50000000 IU/mL |
Pack of 7 for $2,320.00
|
|
||||
Description
Assess the performance of molecular test procedures, including the extraction step, for the quantitative and qualitative determination of HIV-1, HCV and HBV nucleic acids with the Thermo Scientific™ AcroMetrix™ HIV-1/HCV/HBV Panels (RUO). These panels can be used for training and proficiency testing of laboratory personnel, for lot-to-lot comparison of reagent test kits and to evaluate and compare intralaboratory and interlaboratory assessments. The panels are for Research Use Only and are not for use in diagnostic procedures.
Specifications
7 x 1.2mL | |
7 x 1.2 mL |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.