Learn More
Thermo Scientific™ AcroMetrix™ HIV-1/HCV/HBV Controls (IVD)
Assess the performance of molecular assays that detect human immunodeficiency virus type 1, Hepatitis B and C with bulk controls, which are for In Vitro Diagnostic Use.
$6999.00 - $8062.00
Catalog Number | Mfr. No. | Level | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Level | Price | Quantity & Availability | |||||
23-279-128
|
Thermo Scientific™
967150 |
Low |
Each for $6,999.00
|
|
|||||
23-279-129
|
Thermo Scientific™
967151 |
High |
Each for $8,062.00
|
|
|||||
Description
Assess the performance of molecular assays that detect human immunodeficiency virus type 1, Hepatitis C and Hepatitis B in patient samples with the Thermo Scientific™ AcroMetrix™ HIV-1/HCV/HBV Controls (IVD). Provided in 50mL bulk format and composed of multiple analytes, the AcroMetrix HIV-1/HCV/HBV Controls provide end-user workflow flexibility and cost-efficiency. Manufactured in accordance with ISO 17511, these controls are designed to yield consistent results from lot-to-lot and are ideal for monitoring assay performance over time. The controls are for In Vitro Diagnostic Use.
Specifications
HIV-1/HCV/HBV Control | |
50mL |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.