Learn More
Cytiva AcroPrep™ Advance 96-well Filter Plates for Solvent Filtration - 350 µL, 0.45 µm wwPTFE membrane (10/pkg)
Supplier:  Cytiva 8584
Encompass_Preferred
Robust Chemical Resistance Provides Compatibility with Harsh Organics and Aqueous Solutions•Chemically resistant materials provide a stable platform to process samples in organic solvents•Inert materials of construction ensure complete recovery of samples and low non-specific binding•New well design results in faster, more uniform filtration rates across the plate and reduces hold-up volume•Innovative outlet tip geometry provides direct flow of samples into receiver plate without concerns of cross-contamination This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-206-3160
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.