Learn More
Cytiva AcroPrep™ Advance 96-well 96-well for Nucleic Acid Binding - Long Tip Filter Plate for Nucleic Acid Binding (5/pkg)
Supplier:  Cytiva 8133
Encompass_Preferred
The Pall AcroPrep Advance 96-well Long Tip Filter Plate for Nucleic Acid Binding (NAB) offers researchers the flexibility to purify nucleic acids from a wide variety of starting material types: a single plate for multiple applications. The Pall NAB plate is a multipurpose plate providing flexibility in applications, reduced risk of cross contamination and smooth flow for sample processing.Maximum yields and quality of nucleic acid purification•Outlet tip geometry provides direct flow of samples into receiver plate without concerns of cross contamination•Silica-based quartz glass fiber media that allows efficient binding of DNA and RNA, while providing smooth flow and rapid processing of samples•Manufactured in accordance with ANSI/SBS x-2004 specifications for multi-well plates allowing entire DNA purification process to be performed on automated equipment This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-206-3154
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.