Learn More
Cytiva Ion Chromatography (IC) Acrodisc Syringe Filters - 0.45 µm (1000/pkg)
Supplier:  Cytiva AP4785
Encompass_Preferred
Pall Ion Chromatography (IC) Acrodisc syringe filters have been specifically designed and manufactured for IC applications. They feature an optimized Supor membrane that has undergone several extra washing steps ensuring the product has low conductivity detectable extractables.The syringe filters have ion chromatography certification and are tested using a highly sensitive IC protocol to monitor inorganic extractables and are certified for low levels of inorganic extractables, with actual background levels of filter extractables that are typically less than 20 ppb for chloride, 6 ppb for nitrate, 1 ppb for phosphate, and 10 ppb for sulfate.Polypropylene housing which has excellent chemical compatibility properties. This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-206-3420
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.