Learn More
Electron Microscopy Sciences Aclar Fluoropolymer Film 7.8 Mil, 8x10"
Supplier:  Electron Microscopy Sciences 5042510
Encompass_Preferred
ACLAR is an Embedding/Cell Growing Film that is made from fluorinated-chlorinated resins. The chemical make-up of all ACLAR products provide an exceptional moisture barrier. ACLAR is crystal clear, biochemically inert, highly resistant to most chemicals and sterilizable by heat or radiation. ACLAR is used widely in pharmaceutical, medical, sensitive electronics and military packaging. ACLAR 33C is a copolymer film consisting primarily of chlorotrifluoroethylene (CTFE) with an outstanding moisture barrier, excellent chemical resistance and minimal dimensional change (<2%). Best choice for microscopy.
Dimensions: 8x10"
Catalog No. 50-997-411
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.