Learn More
Hach Company Acid Reagent Powder Pillows for HR Silica, 10-mL, pk/100
Supplier:  Hach Company 2107469
Encompass_Preferred
For determination of high range silica by the silicomolybdate method (Hach method 8185) Range:1 to 100 mg/L. Sample Size: 10 mL sample. Pack of 100 Powder Pillows.
Catalog No. 50-199-5420
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.