Learn More
Enzo Life Sciences N-Acetyl-S-geranylgeranyl-L-cysteine (25mg). CAS: 139332-94-8
Supplier: Enzo Life Sciences BMLG2200025
AGGC specifically blocks methyl esterification of geranylgeranylated proteins such as Rab proteins. AGGC blocks receptor-mediated signal transduction in human neutrophils (IC50=4 µM). Induces insulin release from HIT-T15 cells by mimicking the action of Rab3A and inhibits β2-integrin-induced actin polymerization (IC50~45 nM) in neutrophils. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (>25mg/ml) or 100% ethanol (>25mg/ml). Long Term Storage: -20°C.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.