Learn More
Supply Solutions Burdick & Jackson - Acetonitrile, BioSyn, Package Size: 56L, Container Type:SSPDS, Suggested Applications: Biosynthesis, Applications requiring low water content
Supplier:  Supply Solutions BJ BC01756
Encompass_Preferred
75-05-8. Acetonitrile, BioSyn. Suggested Application: Biosynthesis, Applications requiring low water content. Package Size: 20L. Container type is steel NOWPAK. Hazard Class: 3. Package Group: II. Molecular Weight: 41.05. Density (g/mL) at 20deg C: 0.7822. Boiling point (deg C): 81.6. Freezing point (deg C): -43.8. Flashpoint (deg C): 6. Manufacturer Name: Burdick and Jackson. Manufacturer Part Number: BN017-20.; Size: 56L (SSPDS)
Catalog No. 50-877-344
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.