Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Specifications
pH Buffer, individual use pouches | |
pH calibration | |
7 | |
Buffer | |
Yes |
pH 7.00 (Yellow) Buffer | |
pH Meter and Electrode, single-use | |
10 x 15 mL | |
Pouch |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Share a Content Correction