Learn More
Andwin Scientific ACCUEDGE BLADE SHARPSHARP 5/PK
SDP
Supplier:  Andwin Scientific 6257005
Encompass_Preferred
The Accu-Edge replacement blade scissors by Sakura Finetek feature sharp blades that also eliminate the chance of cross-contamination every time it is used for tissue processing. The blades are autoclavable and are free of corrosion. Made from stainless steel, the blades can be inserted and removed with the use of an enclosed tool with no difficulty. This product is sold in a pack of 5 pairs of sharp-sharp blades.
Catalog No. 50-727-632
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.