Learn More
Enzo Life Sciences Ac-IETD-pNA (1mg)
SDP
Supplier:  Enzo Life Sciences ALX260045M001
Encompass_Preferred
Caspase and granzyme substrate. Chromogenic substrate for caspase-6, -8 and granzyme B. Also a substrate for caspase-10. Similar to Ac-IETD-AMC and Ac-IETD-AFC but cleavage is monitored colorimetrically by absorbance at 405 nm. Alternative name: Caspase-8 substrate (chromogenic), Granzyme B substrate (chromogenic). Formula: C27H38N6O12. MW: 638.6. Purity: ≥98% (HPLC). Formulation: Lyophilized. Peptide content: 70-90%. Solubility: Soluble in DMSO. Dilute with buffer, pH 7.0-7.5. Long Term Storage: -20°C. Handling: Keep cool and dry.
Catalog No. 50-200-8256
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.