Learn More
Enzo Life Sciences Ac-DEVD-AMC (5 mg)
SDP
Supplier:  Enzo Life Sciences ALX260031M005
Encompass_Preferred
Caspase substrate. Fluorogenic substrate for caspase-3 with Km=9.7µM. Also a substrate for caspase-1, -4, -7 and -8. The peptide sequence is based on the PARP cleavage site Asp216 for caspase-3. Ex.: 340-360nm, Em.: 440-460nm. Alternative Name: Caspase 3 substrate (fluorogenic). Formula: C30H37N5O13. MW: 675.7. Formulation: Lyophilized. Purity: ≥97% (HPLC). Sequence Ac-Asp-Glu-Val-Asp-AMC (AMC = 7-Amino-4-methylcoumarin). Solubility: Soluble in DMSO; dilute with buffer, pH 7.5. Long Term Storage: -20°C. Handling: Protect from light.
Catalog No. NC0550669
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.