Learn More
Cole-Parmer Megger 230425 AC/DC Hi-Pot Tester, 0/4kV 5kVDC
Supplier:  Cole-Parmer 2002265
Encompass_Preferred
These AC and AC/DC High-Pot Testers are designed for safety and ease of operation, and readily adapt to development, maintenance or production testing. Three models are available: two AC and one AC/DC. The AC High-Pot Testers feature a front-panel, NEMA 5-15IR three-prong output receptacle. Booted, alligator-clip output and return probes are also supplied with all models.Test voltage is continuously adjustable from zero to full output, with a zero-start interlock. Leakage current trip level may be adjusted from 0.3 to 12 mA by a front-panel knob. Push buttons are provided to select one-second or continuous testing, to turn off the high voltage and to reset the instrument. All high-voltage connections are recessed to prevent accidental contact with high voltage.
Catalog No. 50-993-496
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.