Learn More
Carbosynth LLC. Ac-cyclo [Pen-Tyr(Me)-Ala-Arg-Gly-Asp-Asn-Tic-Cys]-NH2, 1mg, MF= C142H234N36O35S3
SDP
Supplier:  Carbosynth LLC. RGD3035PI1MG
Encompass_Preferred
Application: Cell Penetrating Peptide That Targets EGFR, Binds to a Specific Protein in Carcinoma Cells, and Induces Apoptosis, salt: Trifluoroacetate Salt, MW: 3101.86
Catalog No. 50-195-2427
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.