Learn More
Carbosynth LLC. Ac-cyclo [Pen-Tyr(Me)-Ala-Arg-Gly-Asp-Asn-Tic-Cys]-NH2, 5mg, MF= C142H234N36O35S3
Supplier: Carbosynth LLC. RGD3035PI5MG
Application: Cell Penetrating Peptide That Targets EGFR, Binds to a Specific Protein in Carcinoma Cells, and Induces Apoptosis, salt: Trifluoroacetate Salt, MW: 3101.86