Learn More
Enzo Life Sciences Ac-AAVALLPAVLLALLAPYVAD-CHO (1mg)
SDP
Supplier:  Enzo Life Sciences ALX260047M001
Encompass_Preferred
Caspase-1 inhibitor. The YVAD-CHO portion of this peptide is a highly-specific, potent (Ki=1 nM) and reversible inhibitor of caspase-1 (interleukin-1β converting enzyme, ICE), whose sequence is based on the pre-interleukin-1β Asp-116 cleavage site for caspase-1. The sequence linked N-terminal to YVAD-CHO (residues 1-16) has been shown to enhance cell permeability. Alternative name: Caspase 1 inhibitor . Formula: C97H160N20O24. MW: 1990.5. Purity: ≥97% (HPLC). Peptide content: 78-98%. Solubility: Soluble in DMSO (0.25mg/ml). Use/Stability: Preincubate cultured cells 1-2 hrs. with 0.1-2.0µM of the peptide, before inducing for apoptosis. Measure indicator(s) of apoptosis at a number of time points, since the effect of the inhibitor may be a delay in the onset of apoptosis, rather than permanent inhibition. It may also be helpful to determine a minimum effective concentration for the apoptosis inducer and use that concentration, when attempting to observe the effects of this peptide.
Catalog No. 50-200-8260
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.