Learn More
Neogen Diagnostics ABTS Microwell Substrate, 100 ML, HRP, Microwell, Colorimetric
Supplier:  Neogen Diagnostics 310247
Encompass_Preferred
ABTS Substrate is a one-bottle, ready to use substrate for horseradish peroxidase (HRP) microwell applications. It contains 2.2’-azino-bis(3-ethylbenzothiazoline-6-sulfonic acid)-diammonium salt in a mildly acidic buffer. ABTS develops a blue-green color in the presence of a peroxidase labelled conjugate. The reaction can be stopped using 1% SDS stop solution. ABTS substrate is not applicable for use with assays requiring a precipitating substrate. Stable for a minimum of 36-months when stored at 2-8C.
Catalog No. NC2215565
Explore all special offers
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.