Learn More
STEMCELL Technologies ABT-888 (Dihydrochloride), Size: 25 mg
Supplier: STEMCELL Technologies 1001170
Inhibit cancer cell growth by inhibiting poly ADP ribose polymerase (PARP) with ABT-888 (Dihydrochloride). With a high affinity for both PARP1 and PARP2 (IC50 5.2 and 2.9 nM) ABT-888 is inactive against sirtuin 2 (Donawho et al.).