Learn More
Enzo Life Sciences A1874 (10mg). CAS: 2064292-12-0
Supplier: Enzo Life Sciences ENZCHM2620010
A1874 is a nutlin-based PROTAC targeting on BRD4, consist of a BRD4-ligand JQ1 linked to a MDM2 E3 ligase ligand RG7388. A1874 possesses dual function as it dose-dependently degraded its target BRD4, accompanied with the downregulation of c-MYC, as well as stabilization of P53 and upregulated P21 due to the efficient engagement of MDM2 by RG7388. Purity: ≥98%. Long Term Storage: -20°C.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.