Learn More
New England Biolabs, Inc. α1-3,4 Fucosidase - 200 units
SDP
Supplier:  New England Biolabs, Inc. P0769S
Encompass
1-3,4 Fucosidase, clones from almond meal, and also known as AMF, is a broad specificity exoglycosidase that catalyzes the hydrolysis of terminal, non-reducing 1-3 and 1-4 linked fucose residues from oligosaccharides and glycoproteins.
Catalog No. 50-104-7518
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.