Learn More
Bioassay Systems a-Glucosidase Assay Kit
SDP
Supplier:  Bioassay Systems DAGD100
Encompass_Preferred
For quantitative determination of α-glucosidase activity and evaluation of drug effects on its metabolism. Key Features: High sensitivity and wide linear range. Use 20 µL sample. The detection limit is 2 U/L, linear up to 250 U/L. Homogeneous and simple procedure. Simple "mix-and-measure" procedure allows reliable quantitation of α-glucosidase activity within 20 minutes. Robust and amenable to HTS. All reagents are compatible with high-throughput liquid handling instruments. Method: OD405nm. Samples: Biological. Species: All. Procedure: Assay takes 20 min. Kit size: 100 tests. Detection limit: 2 U/L.
Catalog No. 50-225-5859
Explore all special offers
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.