Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

A 412997 dihydrochloride

Selective D4 agonist

$125.40 - $464.00

Specifications

Chemical Name or Material N-(3-Methylphenyl)-4-(2-pyridinyl)-1-piperidineacetamide
CAS 1347744-96-0
Assay Percent Range >98%
Molecular Formula C19H23N3O.2HCl
Formula Weight Observed MW: 382.33
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
45-522-5
View Documents
R&D Systems
4552/25
25 mg
Each for $464.00
Add to Cart
 
45-525
View Documents
R&D Systems
4552/5
5 mg
Each for $125.40
Add to Cart
 
Description

Description

Selective agonist for the dopamine D4 receptor (Ki values are 7.9 and 12.1 nM for human D4 and rat D4 receptors). Displays no affinity (<1000 nM) for other dopamine receptors. Shows efficacy in rat models of ADHD and short-term memory.
Specifications

Specifications

N-(3-Methylphenyl)-4-(2-pyridinyl)-1-piperidineacetamide
>98%
Observed MW: 382.33
1347744-96-0
C19H23N3O.2HCl
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Desiccate at Room Temperature

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.