Learn More
Andwin Scientific 96WPLT PP DW HLF-ML VB CL 80CS
Supplier: Andwin Scientific 786201
The 96-well MASTERBLOCK� microplates is ideal for storing sample materials and cultivating bacteria or yeasts. This product has a high chemical resistance and great temperature endurance ranging from -196�C to 121�C. With alphanumeric well coding, sample identification can be easily done. This product can be sealed with adhesive films and heat sealer and can be used with automated systems.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.