Learn More
Andwin Scientific 96-WELL TEMPPLATE I PP PCR PL
SDP
Supplier:  Andwin Scientific 14029598
Encompass_Preferred
96-well TempPlate;I PP PCR plate, no skirt, low raised rims, mixed colors, PP, 10 plates per pack
Catalog No. 50-819-655
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.