Learn More
Enzo Life Sciences 8(S)-Hydroxyeicosa-5Z,9E,11Z,14Z-tetraenoic acid (HETE) (50µg). CAS: 98462-03-4
SDP
Supplier:  Enzo Life Sciences BMLH0080050
Encompass_Preferred
Biosynthesis in human neutrophils, phorbol ester-treated mouse epidermis and by hepatic microsomal cytochrome P-450. Chemotactic for neutrophils. Activator of PPARα. Activates mouse skin protein kinase C (PKC). Alternative name: 8(S)-HETE. Formulation: Liquid. In ethanol. Purity: ≥99% (HPLC). Long Term Storage: -80°C.
Catalog No. 50-201-1017
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.