Learn More
Enzo Life Sciences 8-Hydroxyguanosine (5mg). CAS: 3868-31-3
SDP
Supplier:  Enzo Life Sciences ALX480075M005
Encompass_Preferred
Oxidized form of the nucleoside guanosine. Formed in vivo by the free radical oxidation of RNA molecules, with subsequent degradation and cleavage of the nucleic acid. Alternative name: 8-OHG. Purity: ≥98% (HPLC). Solubility: Soluble in conc. NaOH. Once solubilized it can be further diluted and neutralized with water. Long Term Storage: -20°C.
Catalog No. 50-200-9317
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.