Learn More
Andwin Scientific 8-CAP STRIPS FOR 1.2 ML MICRO-
SDP
Supplier:  Andwin Scientific 14940400
Encompass_Preferred
The 8-cap strip are compatible with 1.2 mL racked tubes in the 1412-14xx rack series. The strips may also be cut into shorter strips. The caps are free of RNase, DNase, DNA, pyrogen, and PCR inhibitors. It is sold in a pack of 1000 caps.
Catalog No. 50-819-819
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.