Learn More
Andwin Scientific 8-CAP STRIPS FOR 0.2 ML PCR TU
Supplier: Andwin Scientific 14003800
The 8-cap strip is compatible with 0.2 mL PCR tubes and TempPlate 0.2 mL PCR plates. The caps are clear, which allow users to easily observe real-time reactions. It is free of detectable RNase, DNase, DNA, pyrogen, and PCR inhibitors. This product is sold in a pack of 1000 caps.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.