Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
8-Bromo-6-chloroimidazo[1,2-a]pyridine, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
Description
8-Bromo-6-chloroimidazo[1,2-a]pyridine is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
8-Bromo-6-chloroimidazo[1,2-a]pyridine is used as an organic chemical synthesis intermediate.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
8-Bromo-6-chloroimidazo[1,2-a]pyridine is used as an organic chemical synthesis intermediate.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
Specifications
Melting Point | 125°C to 128°C |
Quantity | 250 mg |
Solubility Information | Slightly soluble in water. |
Formula Weight | 231.48 |
Percent Purity | 95% |
Chemical Name or Material | 8-Bromo-6-chloroimidazo[1,2-a]pyridine |
Safety and Handling
Hazard Category | H315-H319-H335 |
Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction