Learn More
Ace Glass, Inc. 7mm OD x 135mm porosity C filter stick
SDP
Supplier:  Ace Glass, Inc. 943523
Encompass_Preferred
7mm O.D. x 135mm gas dispersion tube for fine dispersion. Porous tip is approximately 7mm O.D. x 10mm, and fits a #7 Ace-Thred. Supplied with porosity C (25-50 micron) filter stick.
Catalog No. 50-955-8707
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.