Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ 7X™-PF-O-Matic Detergent

7X™-PF-O-Matic Detergent is a Phosphate Free detergent, specifically developed for safe and effective cleaning of laboratory instruments and glassware.

Supplier:  MP Biomedicals™ 097667593

 View more versions of this product

Catalog No. ICN7667593


Explore all special offers
Add to Cart

Description

Description

  • Environmentally safe and phosphate free
  • Developed for safe and effective cleaning of laboratory instruments and glassware including test tubes, petri-dishes, flasks, microscopy slides, pathology equipment and surgical instruments
  • Powerful wetting and penetrating agent, designed to clean under pressure, heat and agitation
  • Behaves and remains mostly suds-free when agitated
  • Several convenient package sizes are available
  • Stable for long-term storage
  • Completely drains from glassware in seconds
  • Eliminates residue leaving no interfering fluorescence
  • pH neutral, and no need for pH adjustment at any concentration
  • No etch to glass or plastic labware at any concentration
Specifications

Specifications

Liquid
Dark Yellow
15°C to 30°C
1 gal.
7 to 8
7X
Use on Glass, Plastic, automatic washers
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.