Learn More
Sklar Instruments Scissors, Operating Scissors, Sklar 7-1/4 in., Sharp/Blunt, Straight, Smooth, Stainless Steel, Reusable, EA.
Supplier: Sklar Instruments 141072
Scissors, Operating Scissors, Sklar, 7-1/4 in., Sharp/Blunt, Straight, Smooth, Stainless Steel, Reusable, EA. Sklar Operating Scissors, also known as Surgical Scissors, are a versatile surgical instrument. With three available tip configurations (sharp/sharp, blunt/blunt, or sharp/blunt), varying curvatures and multiple lengths, these scissors are used for general, multipurpose cutting and dissecting. Sharp Operating Scissors cut tougher tissue and are frequently used for dissecting, whereas blunt Operating Scissors are more often used for cutting materials such as sutures, tubing, or gauze. This product is straight with smooth, sharp/blunt tips and a length of 7-1/4 inches.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.