Learn More
Chemglass Life Sciences LabJack, Stainless Steel, 15 x 15cm (6 x 6") Top Plate, 25cm (9.8in) Vertical Lift
SDP
Supplier:  Chemglass Life Sciences CG305621
Encompass_Preferred
LabJack, Stainless Steel, 15 x 15cm (6 x 6") Top Plate, 25cm (9.8in) Vertical Lift. Support Jack provide for stable height adjustment and support for items in the lab such as flasks, baths or hot plate stirrers. Top and bottom plates are constructed of stainless steel while the support mechanism is made of chrome plated steel.
Catalog No. 50-390-5
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.