Learn More
Sklar Instruments Scissors, Multi-Cut Utility Scissors, Sklar Black, 6 in., Blunt/Blunt, Angled, Serrated, Stainless Steel / Plastic, EA.
SDP
Supplier:  Sklar Instruments 111279
Encompass_Preferred
Scissors, Multi-Cut Utility Scissors, Sklar, Black, 6 in., Blunt/Blunt, Angled, Serrated, Stainless Steel / Plastic, EA. Sklar Utility Scissors are a multipurpose instrument used for cutting non-sterile materials such as dressings, casts, and surgical drapes. These scissors are angled and have one serrated blade and one smooth blade. The flattened, wide, blunt tip on the serrated blade greatly reduces the likelihood of skin being nicked if the scissors are used for cutting material that is on the patient, such as clothing or bandages. They should never be used to cut tissue or sutures. One plastic finger ring is larger than the other for maximized control while cutting.
Catalog No. 50-109-3312
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.