Learn More
Andwin Scientific 6 X 6 LAB JACK
Supplier: Andwin Scientific 646424
The 6" x 6" Stainless Steel Lab Jack has a platform size of 6" x 6" (150 mm x 150 mm) and an adjustable height of 3"-10". This product is constructed of high quality 304 stainless steel.