Learn More
DBA: SABEU 6 well insert 0.4um PET 4Plate (cellQART)
Encompass_Preferred
cellQART Cell Culture Inserts combined with our especially designed Well Plates are intended to be used for cell culture and in vitro tissue culture research applications. The Well Plate is supplied in 6-Well format, it is sterile and fully pre-loaded with cellQART Cell Culture Inserts. These cellQART Cell Culture Inserts come with PET membranes in clear optics. The Inserts are tissue culture treated and sterile. They are ideal for in-situ phase contrast microscopy.
Catalog No. 50-267-3526
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.