Learn More
Enzo Life Sciences 6-keto-PGF1alpha ELISA kit (5x96wells)
Supplier: Enzo Life Sciences ADI901004
The 6-keto-PGF1α EIA kit is a colorimetric competitive enzyme immunoassay kit with results in < 3 hours. Absorbance is read at 405 nm. This is a highly specific kit with low cross-reactivity to structurally related molecules. The 6-keto-PGF1α EIA kit has been fully validated in-house in a wide variety of sample matrices. Alternative name: 6-keto Prostaglandin F1alpha. Assay Time: <3 hours. Applications: ELISA, Colorimetric detection . Species reactivity: Species independent .
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.