Learn More
Sklar Instruments Scissors, Mayo-Noble Dissecting Scissors, Sklar, 6-1/2 in., Blunt/Blunt, Curved, Smooth, Stainless Steel, EA.
SDP
Supplier:  Sklar Instruments 152862
Encompass_Preferred
Scissors, Mayo-Noble Dissecting Scissors, Sklar, 6-1/2 in., Blunt/Blunt, Curved, Smooth, Stainless Steel , EA. Sklar Mayo-Noble Dissecting Scissors are a slightly heavier variation of the traditional Mayo Dissecting Scissors that still feature the standard Mayo beveled edge. This product is curved with smooth, blunt/blunt tips, and an overall length of 6 1/2 inches.
Catalog No. 50-109-3389
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.