Learn More
Ted Pella Inc Unconjugated Gold Sols - Gold Colloids 5nm | 100mL
SDP
Supplier:  Ted Pella Inc 157021
Encompass
Nanometer-sized gold particles of uniform shape and size are invaluable tools in nanotechnology (assembled arrays), light scattering (elastic - plasmon resonance; inelastic - Raman effect) and for single molecule detection. Gold nanoparticles can be attached to proteins, alkanethiols and DNA by various methods. Silver nanoparticles are evolving similar functional characteristics to their gold counterparts.
Catalog No. NC9007364
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.