Learn More
MTC Bio 5mL Screw-cap MacroTube Vivid™, sterile, w/ vivid printed graduation w/ attached screw caps in foam racks, 500/cs
Supplier: MTC Bio C2545
MTC Bio's MacroTube Vivid is the only 5mL screw-cap tube with printed graduations and marking area. 5mL Screw-Cap MacroTubes take all of the conveniences of standard 15mL conical tubes and scales them down for space efficiency and decreased natural resource consumption. Useful from -86°C to 110°C, these tubes are a valuable resource for boiling and for sample preservation at very low temperatures. Choose from gamma sterilized racked packaging with attached caps or non-sterile bulk packaging with separate caps. The foam racks are similar to the packaging normally associated with 15mL tubes (upright with caps attached), but they require about 60% less storage space. Compatible with standard equipment. Centrifuge at up to 25,000xg. RNase, DNase & pyrogen free. Unsurpassed Clarity. Dual graduations at every 0.2mL and 0.5mL
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.