Learn More
CELLTREAT Scientific Products 5mL Aspirating Pipet, Bulk Packed, Sterile pipette
Supplier: CELLTREAT Scientific Products 229255
CELLTREAT Scientific Products 5mL Aspirating Pipet. Ideal for aspiration of media and sample waste removal. Supplied without graduations or plugs. Rnase/Dnase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, bulk packed. 25 bulk packed/bag, 200/case.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.