Learn More
Andwin Scientific 50ML BRAND CENT TUBES POLYP
SDP
Supplier:  Andwin Scientific 21008242
Encompass_Preferred
The 50 mL SuperClear™ Ultra-High Performance Centrifuge Tubes feature a thick-wall construction that permits high-speed operation. The tubes are also leakproof, providing a secure storage for samples. This product is free of RNase, DNase, and endotox
Catalog No. NC9947379
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.