Learn More
Chemglass Life Sciences 50L Insulating Jacket
SDP
Supplier:  Chemglass Life Sciences CG10011151
Encompass_Preferred
Similar to CG-10009 but Without the Heating Element. For use as a heat or cold shield on Chemglass jacketed reactors. Insulating jacket is made from a heavy fiberglass fabric on the inside with a silicone-coated fiberglass fabric on the outside. Mantle is slit lengthwise to a hole in the bottom center with lacing closure to accommodate a bottom drain valve.
Catalog No. 50-153-9710
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.