Learn More
Hach Company Sension+ 5070 laboratory platinum conductivity cell for general applications
Supplier:  Hach Company LZW5070.97.0002
Encompass_Preferred
Sension+ 5070 is a three-pole platinum conductivity cell with a glass body and built-in temperature sensor. The 5070 has a fixed 1 meter cable and telephonic connector dedicated for use with Hach Sension+ Laboratory Conductivity meters. The 5070 is ideal for conductivity measurements in general aqueous applications.
Catalog No. 50-199-7977
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.